ENAH2C557099 Enamine

6-(methoxycarbonyl)-4-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD31666949
  • Cas Number : 2241130-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008497
Availability details for
Quantity
  • Availability
  • Add to Cart