ENAH2C55709C Enamine

3-(1-phenylethyl)pyrrolidine hydrochloride

  • MDL number MFCD31667070
  • Cas Number : 2230807-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008531
Availability details for
Quantity
  • Availability
  • Add to Cart