ENAH2C5570A4 Enamine

4-(butan-2-yl)-1,3-thiazol-2-amine hydrobromide

  • MDL number MFCD31667119
  • Cas Number : 2230807-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008885
Availability details for
Quantity
  • Availability
  • Add to Cart