ENAH2C5570A6 Enamine

tert-butyl (4aS,7aS)-1-(chlorosulfonyl)-octahydro-1H-pyrrolo[3,4-b]pyridine-6-carboxylate

  • MDL number MFCD31666950
  • Cas Number : 2241107-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008915
Availability details for
Quantity
  • Availability
  • Add to Cart