ENAH2C5570A9 Enamine

4-chloro-3-hydroxy-2,3-dihydro-1lambda6-benzothiophene-1,1-dione

  • MDL number MFCD31667123
  • Cas Number : 2230802-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008937
Availability details for
Quantity
  • Availability
  • Add to Cart