ENAH2C5570AB Enamine

7-thia-2-azaspiro[3.5]nonane hydrochloride

  • MDL number MFCD31667838
  • Cas Number : 2230807-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008961
Availability details for
Quantity
  • Availability
  • Add to Cart