ENAH2C5570AD Enamine

tert-butyl 6-(2,2,2-trifluoroacetyl)-3,4-dihydro-2H-oxazine-4-carboxylate

  • MDL number MFCD31691381
  • Cas Number : 2230799-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008975
Availability details for
Quantity
  • Availability
  • Add to Cart