ENAH2C5570B2 Enamine

4-(aminomethyl)-5-methyl-2H-1,3-dioxol-2-one hydrochloride

  • MDL number MFCD31667842
  • Cas Number : 2230804-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009017
Availability details for
Quantity
  • Availability
  • Add to Cart