ENAH2C5570B8 Enamine

7-chloro-5-(2-fluorophenyl)-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine

  • MDL number MFCD31666952
  • Cas Number : 2241142-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009053
Availability details for
Quantity
  • Availability
  • Add to Cart