ENAH2C5570BB Enamine

2,5-dichloro-5,6,7,8-tetrahydroquinoxaline

  • MDL number MFCD31691044
  • Cas Number : 2230807-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009064
Availability details for
Quantity
  • Availability
  • Add to Cart