ENAH2C5570BF Enamine

5-[(tert-butoxy)carbonyl]-1,1-difluoro-5-azaspiro[2.3]hexane-4-carboxylic acid

  • MDL number MFCD31667846
  • Cas Number : 2230803-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009106
Availability details for
Quantity
  • Availability
  • Add to Cart