ENAH2C5570C1 Enamine

6-oxabicyclo[3.2.1]octane-5-carboxylic acid

  • MDL number MFCD31691384
  • Cas Number : 2230807-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009121
Availability details for
Quantity
  • Availability
  • Add to Cart