ENAH2C5570C5 Enamine

4-(4-bromo-2,3-difluorophenyl)-2,6-dichloropyrimidine

  • MDL number MFCD31691066
  • Cas Number : 2023787-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009187
Availability details for
Quantity
  • Availability
  • Add to Cart