ENAH2C5570C6 Enamine

methyl 2-(carbonochloridoyl)-1,2-oxazolidine-5-carboxylate

  • MDL number MFCD31667243
  • Cas Number : 2230798-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009226
Availability details for
Quantity
  • Availability
  • Add to Cart