ENAH2C5570C8 Enamine

tert-butyl 1-amino-1H-pyrazole-4-carboxylate

  • MDL number MFCD31666953
  • Cas Number : 2241131-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009234
Availability details for
Quantity
  • Availability
  • Add to Cart