ENAH2C5570CD Enamine

tert-butyl 2-(prop-2-yn-1-yl)oxolane-2-carboxylate

  • MDL number MFCD31691385
  • Cas Number : 2230807-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009277
Availability details for
Quantity
  • Availability
  • Add to Cart