ENAH2C5570D1 Enamine

[2-methanesulfonyl-6-(2,2,2-trifluoroethoxy)phenyl]methanol

  • MDL number MFCD31667929
  • Cas Number : 2230800-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009294
Availability details for
Quantity
  • Availability
  • Add to Cart