ENAH2C5570D6 Enamine

{2-[(2S)-4-methylmorpholin-2-yl]pyrimidin-4-yl}methanol

  • MDL number MFCD31691072
  • Cas Number : 2230789-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009372
Availability details for
Quantity
  • Availability
  • Add to Cart