ENAH2C5570D7 Enamine

3-(2-aminoethyl)-5,5-dimethylpyrrolidin-2-one hydrochloride

  • MDL number MFCD31667931
  • Cas Number : 2230804-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009377
Availability details for
Quantity
  • Availability
  • Add to Cart