ENAH2C5570DA Enamine

4-(3-chloropropyl)-1,4-oxazepane hydrochloride

  • MDL number MFCD31667249
  • Cas Number : 2230803-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009388
Availability details for
Quantity
  • Availability
  • Add to Cart