ENAH2C5570DC Enamine

(1s,3s)-3-(5-amino-1-methyl-1H-1,2,4-triazol-3-yl)cyclobutan-1-ol

  • Cas Number : 2230789-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009403
Availability details for
Quantity
  • Availability
  • Add to Cart