ENAH2C5570DF Enamine

1-(carbonochloridoyl)azetidin-3-yl acetate

  • MDL number MFCD31666941
  • Cas Number : 2241128-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009471
Availability details for
Quantity
  • Availability
  • Add to Cart