ENAH2C5570E1 Enamine

3-[(2R)-1-[(tert-butoxy)carbonyl]-2-methylpyrrolidin-2-yl]prop-2-ynoic acid

  • MDL number MFCD31667936
  • Cas Number : 2230789-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009509
Availability details for
Quantity
  • Availability
  • Add to Cart