ENAH2C5570E4 Enamine

2-(4-methylmorpholin-3-yl)pyrimidine-4-carboxylic acid hydrochloride

  • MDL number MFCD31667297
  • Cas Number : 2230803-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009544
Availability details for
Quantity
  • Availability
  • Add to Cart