ENAH2C5570F1 Enamine

tert-butyl 4-[3-(aminomethyl)-3-methyl-2-oxoazetidin-1-yl]piperidine-1-carboxylate

  • MDL number MFCD31667016
  • Cas Number : 2241130-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009746
Availability details for
Quantity
  • Availability
  • Add to Cart