ENAH2C5570F2 Enamine

2-bromo-5,7-dimethoxy-[1,3]thiazolo[4,5-d]pyrimidine

  • MDL number MFCD31668722
  • Cas Number : 2243504-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009748
Availability details for
Quantity
  • Availability
  • Add to Cart