ENAH2C5570F3 Enamine

5,7-dimethoxy-[1,3]thiazolo[4,5-d]pyrimidine

  • MDL number MFCD31668664
  • Cas Number : 2243510-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009749
Availability details for
Quantity
  • Availability
  • Add to Cart