ENAH2C5570F8 Enamine

3-(2,2-difluoroethyl)pyrrolidine hydrochloride

  • MDL number MFCD31667322
  • Cas Number : 2230803-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2009789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2009789
Availability details for
Quantity
  • Availability
  • Add to Cart