ENAH2C5570FC Enamine

2-hydroxy-3-iodo-5-methylbenzoic acid

  • MDL number MFCD30585222
  • Cas Number : 6083-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-220
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722167
Availability details for
Quantity
  • Availability
  • Add to Cart