ENAH2C5570FD Enamine

2-[(2-aminoethyl)amino]ethane-1-thiol

  • MDL number MFCD19203669
  • Cas Number : 51896-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722200
Availability details for
Quantity
  • Availability
  • Add to Cart