ENAH2C557127 Enamine

methyl 4-bromo-2,3,6-trifluorobenzoate

  • MDL number MFCD06204171
  • Cas Number : 773134-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1722913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1722913
Availability details for
Quantity
  • Availability
  • Add to Cart