ENAH2C557137 Enamine

(2R)-2-(4-fluorophenyl)propanoic acid

  • MDL number MFCD18434514
  • Cas Number : 150360-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717377

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-1717377
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart