ENAH2C55713F Enamine

methyl 2-(3-bromophenyl)-2-diazoacetate

  • MDL number MFCD31691280
  • Cas Number : 264882-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723432
Availability details for
Quantity
  • Availability
  • Add to Cart