ENAH2C557140 Enamine

methyl 2-(2-bromophenyl)-2-diazoacetate

  • MDL number MFCD31691399
  • Cas Number : 264882-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723434
Availability details for
Quantity
  • Availability
  • Add to Cart