ENAH2C557143 Enamine

3-(2-aminoethyl)-1-(2,3-dihydro-1H-inden-1-yl)thiourea

  • MDL number MFCD31668135
  • Cas Number : 2243513-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723520
Availability details for
Quantity
  • Availability
  • Add to Cart