ENAH2C55714B Enamine

N-acetyl-N-methylcarbamoyl chloride

  • MDL number MFCD31667890
  • Cas Number : 21777-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723623
Availability details for
Quantity
  • Availability
  • Add to Cart