ENAH2C55714F Enamine

1H,4H,5H,6H,7H,8H-pyrrolo[3,2-c]azepin-4-one

  • MDL number MFCD20231146
  • Cas Number : 21357-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723667
Availability details for
Quantity
  • Availability
  • Add to Cart