ENAH2C557153 Enamine

2-(3-{[(tert-butoxy)carbonyl]amino}-2-oxopiperidin-1-yl)acetic acid

  • MDL number MFCD12024720
  • Cas Number : 733049-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723715
Availability details for
Quantity
  • Availability
  • Add to Cart