ENAH2C557155 Enamine

ethyl 2,4-dimethylimidazo[1,5-a]pyrimidine-8-carboxylate

  • MDL number MFCD29069913
  • Cas Number : 32704-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723741
Availability details for
Quantity
  • Availability
  • Add to Cart