ENAH2C557157 Enamine

methyl 2-chloro-2-(3-fluorophenyl)acetate

  • MDL number MFCD16694215
  • Cas Number : 1249616-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723768
Availability details for
Quantity
  • Availability
  • Add to Cart