ENAH2C55715A Enamine

3-methyl-1H-pyrazolo[3,4-c]pyridine-5-carboxylic acid

  • MDL number MFCD30490068
  • Cas Number : 1501435-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723894
Availability details for
Quantity
  • Availability
  • Add to Cart