ENAH2C557162 Enamine

4-(2-bromoacetyl)-1lambda6-thiomorpholine-1,1-dione

  • MDL number MFCD03840693
  • Cas Number : 887571-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723966
Availability details for
Quantity
  • Availability
  • Add to Cart