ENAH2C557164 Enamine

2-(3-bromoprop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD31667891
  • Cas Number : 574011-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1723995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1723995
Availability details for
Quantity
  • Availability
  • Add to Cart