ENAH2C557166 Enamine

2,4-dichloro-5-(propan-2-yl)pyridine

  • MDL number MFCD28054791
  • Cas Number : 1622840-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724043
Availability details for
Quantity
  • Availability
  • Add to Cart