ENAH2C557167 Enamine

2-bromo-6-fluoro-4-nitrobenzonitrile

  • MDL number MFCD28740043
  • Cas Number : 1807071-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724045
Availability details for
Quantity
  • Availability
  • Add to Cart