ENAH2C557168 Enamine

N-(piperidin-4-yl)pyridin-4-amine

  • MDL number MFCD09933492
  • Cas Number : 181258-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724085
Availability details for
Quantity
  • Availability
  • Add to Cart