ENAH2C55716A Enamine

3,6-dimethyl-1H-indole

  • MDL number MFCD19217438
  • Cas Number : 81784-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724151
Availability details for
Quantity
  • Availability
  • Add to Cart