ENAH2C55716B Enamine

6-fluoro-3-methyl-1H-indole

  • MDL number MFCD03095358
  • Cas Number : 1011484-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724152
Availability details for
Quantity
  • Availability
  • Add to Cart