ENAH2C55716E Enamine

4-nitro-1,3-benzothiazol-5-amine hydrochloride

  • MDL number MFCD31667669
  • Cas Number : 2230808-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724225
Availability details for
Quantity
  • Availability
  • Add to Cart