ENAH2C557173 Enamine

{[(2-bromoprop-2-en-1-yl)oxy]methyl}benzene

  • MDL number MFCD00995640
  • Cas Number : 79629-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1724754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1724754
Availability details for
Quantity
  • Availability
  • Add to Cart